The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000226.3 | NP_000217.2 | 3 - 4 | 954 | 76 |
GenBank mRNA | AB001594.1 | - | 3 - 4 | 451 | 76 |
BC121170.2 | - | 4 - 5 | 420 | 76 | |
S69510.1 | - | 3 - 4 | 951 | 76 | |
Z29074.1 | - | 3 - 4 | 954 | 76 |