The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175921.5 | NP_787117.3 | 5 - 6 | 624 | 135 |
XM_011514032.2 | XP_011512334.1 | 6 - 7 | 753 | 135 | |
GenBank mRNA | BF439305.1 | - | 5 - 6 | 588 | 135 |