Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAPTM4B
Entrez Gene ID
55353
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
LAPTM4beta, LC27
Chromosome Location
Chr.8: 97775581 - 97852602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018407.4 NP_060877.3 3 - 4 720 100
GenBank mRNA AF317417.1 - 3 - 4 291 100
AF527412.1 - 3 - 4 649 100
AK075326.1 - 3 - 4 482 100
AL136942.1 - 3 - 4 474 100
AY057051.2 - 3 - 4 720 100
AY261384.1 - 3 - 4 739 100
BC014129.2 - 3 - 4 476 100
BC031021.1 - 3 - 4 501 100
BC038117.1 - 3 - 4 1268 100

View Full Product Details