Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SF3A1
Entrez Gene ID
10291
Gene Name
splicing factor 3a subunit 1
Gene Aliases
PRP21, PRPF21, SAP114, SF3A120
Chromosome Location
Chr.22: 30331988 - 30356947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005877.5 NP_005868.1 4 - 5 805 96
GenBank mRNA AK303273.1 - 5 - 6 730 96
BC001976.1 - 4 - 5 745 96
BC007684.2 - 4 - 5 772 96
BI827130.1 - 4 - 5 564 96
CR456575.1 - 4 - 5 748 96
X85237.1 - 4 - 5 747 96

View Full Product Details