Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBT
Entrez Gene ID
1629
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B
Chromosome Location
Chr.1: 100186922 - 100249864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001918.3 NP_001909.3 7 - 8 977 92
XM_005270545.3 XP_005270602.1 8 - 9 1175 92
XM_017000468.1 XP_016855957.1 9 - 10 1204 92
XM_017000469.1 XP_016855958.1 5 - 6 632 92
GenBank mRNA AK303941.1 - 8 - 9 1148 92
AK313191.1 - 7 - 8 959 92
BC016675.1 - 7 - 8 944 92
BC030002.1 - 7 - 8 944 92
BT007372.1 - 7 - 8 944 92
J03208.1 - 7 - 8 942 92
M27093.1 - 7 - 8 942 92
X66785.1 - 7 - 8 958 92

View Full Product Details