Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DBT
Entrez Gene ID
1629
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B
Chromosome Location
Chr.1: 100186922 - 100249864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001918.3 NP_001909.3 9 - 10 1247 86
XM_005270545.3 XP_005270602.1 10 - 11 1445 86
XM_017000468.1 XP_016855957.1 11 - 12 1474 86
XM_017000469.1 XP_016855958.1 7 - 8 902 86
GenBank mRNA AK303941.1 - 10 - 11 1418 86
AK313191.1 - 9 - 10 1229 86
BC016675.1 - 9 - 10 1214 86
BC030002.1 - 9 - 10 1214 86
BT007372.1 - 9 - 10 1214 86
J03208.1 - 9 - 10 1212 86
M27093.1 - 9 - 10 1212 86
X66785.1 - 9 - 10 1228 86

View Full Product Details