The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324255.1 | NP_001311184.1 | 2 - 3 | 370 | 71 |
NM_005909.4 | NP_005900.2 | 3 - 4 | 640 | 71 | |
GenBank mRNA | AK096273.1 | - | 4 - 5 | 463 | 71 |
AK307990.1 | - | 2 - 3 | 370 | 71 | |
AK308147.1 | - | 3 - 4 | 431 | 71 | |
BC017240.2 | - | 3 - 4 | 617 | 71 | |
BC025240.1 | - | 3 - 4 | 559 | 71 | |
BC032516.1 | - | 3 - 4 | 432 | 71 | |
BC039822.1 | - | 3 - 4 | 426 | 71 | |
BC046114.1 | - | 3 - 4 | 446 | 71 | |
BC056145.1 | - | 3 - 4 | 420 | 71 | |
BC063669.1 | - | 3 - 4 | 420 | 71 | |
BC073993.1 | - | 3 - 4 | 422 | 71 | |
BC094834.1 | - | 3 - 4 | 417 | 71 | |
BC108733.1 | - | 3 - 4 | 435 | 71 | |
BC139918.1 | - | 3 - 4 | 537 | 71 | |
BC141853.1 | - | 2 - 3 | 223 | 71 | |
BC150196.1 | - | 3 - 4 | 537 | 71 | |
BX647077.1 | - | 3 - 4 | 431 | 71 |