Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC26A10
Entrez Gene ID
65012
Gene Name
solute carrier family 26 member 10
Gene Aliases
-
Chromosome Location
Chr.12: 57619910 - 57626151 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133489.2 NP_597996.2 11 - 12 1686 79
GenBank mRNA AF331523.1 - 11 - 12 1686 79
AK122981.1 - 12 - 13 1354 79
AL050358.1 - 5 - 6 2523 79

View Full Product Details