The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022782.3 | NP_073619.3 | 20 - 21 | 3342 | 119 |
NR_103517.1 | 20 - 21 | 3081 | 119 | ||
XM_011537740.2 | XP_011536042.1 | 20 - 21 | 3246 | 119 | |
XM_011537741.2 | XP_011536043.1 | 20 - 21 | 3246 | 119 | |
XM_017018673.1 | XP_016874162.1 | 20 - 21 | 3340 | 119 | |
XM_017018674.1 | XP_016874163.1 | 19 - 20 | 3144 | 119 | |
XM_017018675.1 | XP_016874164.1 | 19 - 20 | 3148 | 119 | |
XM_017018676.1 | XP_016874165.1 | 19 - 20 | 3090 | 119 | |
XM_017018678.1 | XP_016874167.1 | 19 - 20 | 3066 | 119 | |
XM_017018679.1 | XP_016874168.1 | 21 - 22 | 3279 | 119 | |
XM_017018680.1 | XP_016874169.1 | 19 - 20 | 3188 | 119 | |
XM_017018681.1 | XP_016874170.1 | 19 - 20 | 3101 | 119 | |
GenBank mRNA | AK023016.1 | - | 15 - 16 | 2065 | 119 |
AK091181.1 | - | 16 - 17 | 2671 | 119 | |
AK096541.1 | - | 20 - 21 | 2954 | 119 | |
BC130440.1 | - | 16 - 17 | 2624 | 119 | |
BC144017.1 | - | 15 - 16 | 2276 | 119 | |
X98258.1 | - | 2 - 3 | 124 | 119 |