The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022782.3 | NP_073619.3 | 23 - 24 | 3775 | 80 |
NR_103517.1 | 23 - 24 | 3514 | 80 | ||
XM_017018673.1 | XP_016874162.1 | 23 - 24 | 3773 | 80 | |
XM_017018674.1 | XP_016874163.1 | 22 - 23 | 3577 | 80 | |
XM_017018675.1 | XP_016874164.1 | 22 - 23 | 3581 | 80 | |
XM_017018676.1 | XP_016874165.1 | 22 - 23 | 3523 | 80 | |
XM_017018677.1 | XP_016874166.1 | 21 - 22 | 3454 | 80 | |
XM_017018678.1 | XP_016874167.1 | 22 - 23 | 3499 | 80 | |
XM_017018679.1 | XP_016874168.1 | 24 - 25 | 3712 | 80 | |
XM_017018680.1 | XP_016874169.1 | 22 - 23 | 3621 | 80 | |
XM_017018681.1 | XP_016874170.1 | 22 - 23 | 3534 | 80 | |
GenBank mRNA | AK023016.1 | - | 18 - 19 | 2472 | 80 |
AK091181.1 | - | 19 - 20 | 3104 | 80 | |
AK096541.1 | - | 23 - 24 | 3387 | 80 | |
BC130440.1 | - | 19 - 20 | 3057 | 80 | |
BC144016.1 | - | 17 - 18 | 2827 | 80 | |
BC144017.1 | - | 18 - 19 | 2709 | 80 | |
BF793354.1 | - | 2 - 3 | 213 | 80 | |
BU070541.1 | - | 1 - 2 | 80 | ||
BX092492.1 | - | 2 - 3 | 123 | 80 |