Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POGK
Entrez Gene ID
57645
Gene Name
pogo transposable element with KRAB domain
Gene Aliases
BASS2, KRBOX2, LST003
Chromosome Location
Chr.1: 166839404 - 166856344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017542.4 NP_060012.3 3 - 4 462 79
XM_005245370.3 XP_005245427.1 3 - 4 583 79
XM_011509814.2 XP_011508116.1 3 - 4 739 79
XM_011509815.1 XP_011508117.1 3 - 4 394 79
XM_017001914.1 XP_016857403.1 3 - 4 456 79
GenBank mRNA AB040946.1 - 3 - 4 379 79
AK056711.1 - 3 - 4 360 79
BC131624.1 - 3 - 4 347 79
DB049597.1 - 3 - 4 462 79

View Full Product Details