The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004229.3 | NP_004220.2 | 26 - 27 | 3821 | 136 |
XM_005272701.2 | XP_005272758.1 | 26 - 27 | 3731 | 136 | |
XM_011544000.2 | XP_011542302.1 | 27 - 28 | 3703 | 136 | |
XM_017029962.1 | XP_016885451.1 | 27 - 28 | 3696 | 136 | |
GenBank mRNA | AB006651.1 | - | 26 - 27 | 3800 | 136 |
AF070563.1 | - | 4 - 5 | 669 | 136 | |
AF104256.1 | - | 26 - 27 | 3681 | 136 | |
AF135802.1 | - | 26 - 27 | 3754 | 136 | |
AF304448.1 | - | 26 - 27 | 3800 | 136 | |
AJ012077.1 | - | 2 - 3 | 383 | 136 | |
AK023368.1 | - | 11 - 12 | 1714 | 136 | |
AK293070.1 | - | 26 - 27 | 3999 | 136 | |
BC098377.1 | - | 26 - 27 | 3821 | 136 | |
BC132672.1 | - | 26 - 27 | 3790 | 136 | |
BC132674.1 | - | 26 - 27 | 3790 | 136 |