The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018321.3 | NP_060791.3 | 7 - 8 | 588 | 107 |
GenBank mRNA | AF347667.1 | - | 7 - 8 | 564 | 107 |
AK289610.1 | - | 7 - 8 | 587 | 107 | |
AK307492.1 | - | 3 - 4 | 171 | 107 | |
AY364244.1 | - | 7 - 8 | 564 | 107 | |
BC001546.1 | - | 7 - 8 | 588 | 107 | |
BC006014.1 | - | 7 - 8 | 591 | 107 | |
BC036741.1 | - | 7 - 8 | 938 | 107 |