Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ZNF43
- Entrez Gene ID
-
7594
- Gene Name
- zinc finger protein 43
- Gene Aliases
-
HTF6, KOX27, ZNF39L1
- Chromosome Location
-
Chr.19: 21804946 - 21852095 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DN993158.1 |
- |
1 |
342 |
|
Chr. 19 - 21840844 - 21841443 [-] on Build GRCh38 |
106 |
GenBankmRNA |
AK097763.1 |
- |
1 |
538 |
|
Chr. 19 - 21839597 - 21841647 [-] on Build GRCh38 |
106 |
GenBankmRNA |
BE066012.1 |
- |
1 |
|
|
Chr. 19 - 21841034 - 21841468 [+] on Build GRCh38 |
106 |
GenBankmRNA |
BU187775.1 |
- |
1 |
293 |
|
Chr. 19 - 21840536 - 21841402 [-] on Build GRCh38 |
106 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
zinc finger protein 100 |
ZNF100 |
|
-70.2kb
|
Sense |
Chr. - 21724041 - 21767628 on Build GRCh38 |
Downstream |
zinc finger protein 208 |
ZNF208 |
PMIDP, ZNF95 |
122.8kb
|
Sense |
Chr. - 21966095 - 22010990 on Build GRCh38 |
View Full Product Details