The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005249412.1 | XP_005249469.1 | 1 - 2 | 103 | 87 |
GenBank mRNA | AJ608791.2 | - | 1 - 2 | 264 | 87 |