The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171887.1 | NP_001165358.1 | 4 - 5 | 752 | 127 |
GenBank mRNA | AF161472.1 | - | 4 - 5 | 618 | 127 |
AL117608.1 | - | 2 - 3 | 261 | 127 | |
AY506561.1 | - | 3 - 4 | 399 | 127 | |
BI829470.1 | - | 4 - 5 | 596 | 127 |