Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HMGN3
Entrez Gene ID
9324
Gene Name
high mobility group nucleosomal binding domain 3
Gene Aliases
PNAS-24, PNAS-25, TRIP7
Chromosome Location
Chr.6: 79201245 - 79234738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201362.1 NP_001188291.1 3 - 4 272 75
NM_001201363.1 NP_001188292.1 3 - 4 272 75
NM_001318884.1 NP_001305813.1 3 - 4 272 75
NM_001318886.1 NP_001305815.1 3 - 4 272 75
NM_001318887.1 NP_001305816.1 3 - 4 272 75
NM_001318888.1 NP_001305817.1 3 - 4 272 75
NM_004242.3 NP_004233.1 3 - 4 272 75
NM_138730.2 NP_620058.1 3 - 4 272 75
GenBank mRNA AF274949.1 - 3 - 4 191 75
AF401520.1 - 3 - 4 163 75
AK315391.1 - 3 - 4 216 75
AY043282.1 - 3 - 4 239 75
BC009529.2 - 3 - 4 200 75
BF790582.1 - 3 - 4 168 75
BI551210.1 - 3 - 4 279 75
BM927225.1 - 3 - 4 148 75
CF553005.1 - 3 - 4 136 75
CK023734.1 - 3 - 4 210 75
CX761444.1 - 3 - 4 178 75
DA894213.1 - 3 - 4 216 75
L40357.1 - 3 - 4 157 75

View Full Product Details