Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF2A
Entrez Gene ID
3796
Gene Name
kinesin family member 2A
Gene Aliases
CDCBM3, HK2, KIF2
Chromosome Location
Chr.5: 62306162 - 62387184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098511.2 NP_001091981.1 1 - 2 375 141
NM_001243953.1 NP_001230882.1 1 - 2 375 141
NM_004520.4 NP_004511.2 1 - 2 375 141
GenBank mRNA AK297347.1 - 1 - 2 121 141
AK307880.1 - 1 - 2 85 141
AK315866.1 - 1 - 2 59 141
DB452624.1 - 1 - 2 294 141
EF560716.1 - 1 - 2 86 141
EF560728.1 - 1 - 2 80 141

View Full Product Details