The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016441.2 | NP_057525.1 | 14 - 15 | 2688 | 71 |
XM_005264357.4 | XP_005264414.1 | 13 - 14 | 2510 | 71 | |
XM_011532898.2 | XP_011531200.1 | 15 - 16 | 2809 | 71 | |
XM_011532901.2 | XP_011531203.1 | 14 - 15 | 2588 | 71 | |
XM_017004258.1 | XP_016859747.1 | 14 - 15 | 2566 | 71 | |
XM_017004260.1 | XP_016859749.1 | 13 - 14 | 2441 | 71 | |
XM_017004261.1 | XP_016859750.1 | 13 - 14 | 2101 | 71 | |
XM_017004262.1 | XP_016859751.1 | 12 - 13 | 1984 | 71 | |
GenBank mRNA | AB209139.1 | - | 14 - 15 | 2688 | 71 |
AF167706.1 | - | 14 - 15 | 2661 | 71 | |
AK300817.1 | - | 3 - 4 | 490 | 71 | |
AY358371.1 | - | 14 - 15 | 2989 | 71 | |
BC111989.1 | - | 14 - 15 | 2659 | 71 | |
BC113371.1 | - | 14 - 15 | 2659 | 71 | |
BX648755.1 | - | 8 - 9 | 1337 | 71 |