The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016441.2 | NP_057525.1 | 15 - 16 | 2813 | 83 |
XM_005264357.4 | XP_005264414.1 | 14 - 15 | 2635 | 83 | |
XM_011532898.2 | XP_011531200.1 | 16 - 17 | 2934 | 83 | |
XM_011532899.2 | XP_011531201.1 | 15 - 16 | 2740 | 83 | |
XM_011532901.2 | XP_011531203.1 | 15 - 16 | 2713 | 83 | |
XM_017004258.1 | XP_016859747.1 | 15 - 16 | 2691 | 83 | |
XM_017004259.1 | XP_016859748.1 | 14 - 15 | 2757 | 83 | |
XM_017004260.1 | XP_016859749.1 | 14 - 15 | 2566 | 83 | |
XM_017004261.1 | XP_016859750.1 | 14 - 15 | 2226 | 83 | |
XM_017004262.1 | XP_016859751.1 | 13 - 14 | 2109 | 83 | |
GenBank mRNA | AB209139.1 | - | 15 - 16 | 2813 | 83 |
AF167706.1 | - | 15 - 16 | 2786 | 83 | |
AK300817.1 | - | 4 - 5 | 615 | 83 | |
AY358371.1 | - | 15 - 16 | 3114 | 83 | |
BC111989.1 | - | 15 - 16 | 2784 | 83 | |
BC113371.1 | - | 15 - 16 | 2784 | 83 | |
BX648755.1 | - | 9 - 10 | 1462 | 83 |