The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193308.1 | NP_001180237.1 | 8 - 9 | 943 | 67 |
NM_032872.2 | NP_116261.1 | 8 - 9 | 907 | 67 | |
XM_005246022.3 | XP_005246079.1 | 8 - 9 | 1098 | 67 | |
XM_006710990.1 | XP_006711053.1 | 8 - 9 | 1062 | 67 | |
GenBank mRNA | AK027902.1 | - | 8 - 9 | 877 | 67 |
AK096437.1 | - | 7 - 8 | 1248 | 67 | |
AK293038.1 | - | 7 - 8 | 1209 | 67 | |
AY037157.1 | - | 8 - 9 | 870 | 67 | |
BC009224.2 | - | 6 - 7 | 474 | 67 | |
BC015764.1 | - | 7 - 8 | 686 | 67 | |
BC035725.1 | - | 8 - 9 | 850 | 67 |