The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320821.1 | NP_001307750.1 | 15 - 16 | 1884 | 67 |
NM_001320822.1 | NP_001307751.1 | 13 - 14 | 1406 | 67 | |
NM_007368.3 | NP_031394.2 | 13 - 14 | 1418 | 67 | |
XM_011534841.2 | XP_011533143.1 | 13 - 14 | 1483 | 67 | |
XM_011534842.2 | XP_011533144.1 | 13 - 14 | 1543 | 67 | |
XM_017020438.1 | XP_016875927.1 | 15 - 16 | 1832 | 67 | |
XM_017020439.1 | XP_016875928.1 | 14 - 15 | 1629 | 67 | |
GenBank mRNA | AK128094.1 | - | 15 - 16 | 1853 | 67 |
AK289746.1 | - | 13 - 14 | 1324 | 67 | |
AK294913.1 | - | 13 - 14 | 1391 | 67 | |
AK313795.1 | - | 13 - 14 | 1352 | 67 | |
BC038456.1 | - | 13 - 14 | 1415 | 67 | |
BC047242.1 | - | 13 - 14 | 1406 | 67 | |
X89399.1 | - | 13 - 14 | 1330 | 67 |