The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320821.1 | NP_001307750.1 | 6 - 7 | 969 | 82 |
NM_001320822.1 | NP_001307751.1 | 4 - 5 | 491 | 82 | |
NM_007368.3 | NP_031394.2 | 4 - 5 | 503 | 82 | |
XM_011534841.2 | XP_011533143.1 | 4 - 5 | 568 | 82 | |
XM_011534842.2 | XP_011533144.1 | 4 - 5 | 628 | 82 | |
XM_017020438.1 | XP_016875927.1 | 6 - 7 | 917 | 82 | |
XM_017020439.1 | XP_016875928.1 | 5 - 6 | 714 | 82 | |
GenBank mRNA | AK128094.1 | - | 6 - 7 | 938 | 82 |
AK289746.1 | - | 4 - 5 | 409 | 82 | |
AK294913.1 | - | 4 - 5 | 476 | 82 | |
AK313795.1 | - | 4 - 5 | 437 | 82 | |
BC038456.1 | - | 4 - 5 | 500 | 82 | |
BC047242.1 | - | 4 - 5 | 491 | 82 | |
DA328760.1 | - | 4 - 5 | 504 | 82 | |
X89399.1 | - | 4 - 5 | 415 | 82 |