The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320821.1 | NP_001307750.1 | 11 - 12 | 1381 | 99 |
NM_001320822.1 | NP_001307751.1 | 9 - 10 | 903 | 99 | |
NM_007368.3 | NP_031394.2 | 9 - 10 | 915 | 99 | |
XM_011534841.2 | XP_011533143.1 | 9 - 10 | 980 | 99 | |
XM_011534842.2 | XP_011533144.1 | 9 - 10 | 1040 | 99 | |
XM_017020438.1 | XP_016875927.1 | 11 - 12 | 1329 | 99 | |
XM_017020439.1 | XP_016875928.1 | 10 - 11 | 1126 | 99 | |
GenBank mRNA | AK128094.1 | - | 11 - 12 | 1350 | 99 |
AK289746.1 | - | 9 - 10 | 821 | 99 | |
AK294913.1 | - | 9 - 10 | 888 | 99 | |
AK295861.1 | - | 8 - 9 | 749 | 99 | |
AK313795.1 | - | 9 - 10 | 849 | 99 | |
BC038456.1 | - | 9 - 10 | 912 | 99 | |
BC047242.1 | - | 9 - 10 | 903 | 99 | |
X89399.1 | - | 9 - 10 | 827 | 99 |