The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006149.3 | NP_006140.1 | 3 - 4 | 550 | 77 |
XM_011526973.1 | XP_011525275.1 | 3 - 4 | 994 | 77 | |
GenBank mRNA | AB006781.1 | - | 3 - 4 | 392 | 77 |
AF014838.1 | - | 3 - 4 | 349 | 77 | |
AK290269.1 | - | 3 - 4 | 395 | 77 | |
AK296194.1 | - | 3 - 4 | 331 | 77 | |
AK296604.1 | - | 3 - 4 | 395 | 77 | |
BC003661.2 | - | 3 - 4 | 379 | 77 | |
BC005146.2 | - | 3 - 4 | 379 | 77 | |
BC034750.1 | - | 3 - 4 | 375 | 77 | |
BT007032.1 | - | 3 - 4 | 336 | 77 | |
CR536544.1 | - | 3 - 4 | 336 | 77 | |
U82953.1 | - | 3 - 4 | 359 | 77 |