The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000640.2 | NP_000631.1 | 6 - 7 | 826 | 132 |
GenBank mRNA | AK291957.1 | - | 6 - 7 | 795 | 132 |
AK308523.1 | - | 6 - 7 | 757 | 132 | |
AK309444.1 | - | 6 - 7 | 836 | 132 | |
BC020739.1 | - | 5 - 6 | 727 | 132 | |
BC033705.2 | - | 5 - 6 | 735 | 132 | |
GQ494004.1 | - | 6 - 7 | 758 | 132 | |
U70981.1 | - | 6 - 7 | 794 | 132 | |
X95302.1 | - | 6 - 7 | 753 | 132 | |
Y08768.1 | - | 6 - 7 | 826 | 132 |