The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013320.2 | NP_037452.1 | 14 - 15 | 2161 | 70 |
XM_017019241.1 | XP_016874730.1 | 13 - 14 | 1993 | 70 | |
GenBank mRNA | AF117210.1 | - | 14 - 15 | 2070 | 70 |
AK223342.1 | - | 14 - 15 | 2088 | 70 | |
AK313466.1 | - | 14 - 15 | 2091 | 70 | |
BC006558.2 | - | 13 - 14 | 2024 | 70 | |
BC033799.1 | - | 14 - 15 | 2063 | 70 |