The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007086.3 | NP_009017.1 | 2 - 3 | 174 | 67 |
XM_006720012.2 | XP_006720075.1 | 2 - 3 | 172 | 67 | |
GenBank mRNA | AJ006266.1 | - | 1 - 2 | 123 | 67 |
AK293005.1 | - | 2 - 3 | 141 | 67 | |
BC000622.2 | - | 2 - 3 | 139 | 67 | |
BC005040.1 | - | 2 - 3 | 139 | 67 | |
BC043349.1 | - | 2 - 3 | 162 | 67 | |
BC063041.1 | - | 2 - 3 | 150 | 67 | |
DB082418.1 | - | 2 - 3 | 174 | 67 |