The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008396.2 | NP_001008397.1 | 20 - 21 | 2697 | 91 |
NM_007086.3 | NP_009017.1 | 21 - 22 | 2790 | 91 | |
XM_011536373.2 | XP_011534675.1 | 20 - 21 | 2589 | 91 | |
GenBank mRNA | AJ006266.1 | - | 20 - 21 | 2739 | 91 |
AK001538.1 | - | 9 - 10 | 1344 | 91 | |
AK001585.1 | - | 11 - 12 | 1713 | 91 | |
AK001620.1 | - | 4 - 5 | 517 | 91 | |
AK293005.1 | - | 21 - 22 | 2757 | 91 | |
BC063041.1 | - | 21 - 22 | 2766 | 91 |