The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004457.3 | NP_004448.2 | 3 - 4 | 414 | 100 |
GenBank mRNA | AB061712.1 | - | 3 - 4 | 382 | 100 |
AK314621.1 | - | 3 - 4 | 392 | 100 | |
BC041692.1 | - | 3 - 4 | 406 | 100 | |
BG772246.1 | - | 3 - 4 | 499 | 100 | |
BQ435549.1 | - | 3 - 4 | 100 | ||
D89053.1 | - | 1 - 2 | 105 | 100 |