The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004457.3 | NP_004448.2 | 10 - 11 | 1604 | 116 |
NM_203372.1 | NP_976251.1 | 9 - 10 | 1497 | 116 | |
XM_017003584.1 | XP_016859073.1 | 8 - 9 | 1416 | 116 | |
GenBank mRNA | AB061712.1 | - | 10 - 11 | 1572 | 116 |
AK001471.1 | - | 5 - 6 | 523 | 116 | |
AK023191.1 | - | 7 - 8 | 1149 | 116 | |
AK314621.1 | - | 10 - 11 | 1582 | 116 | |
BC041692.1 | - | 10 - 11 | 1596 | 116 | |
D89053.1 | - | 8 - 9 | 1295 | 116 |