The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005065.5 | NP_005056.3 | 12 - 13 | 1377 | 112 |
XM_005267988.2 | XP_005268045.1 | 12 - 13 | 1260 | 112 | |
GenBank mRNA | AB020335.1 | - | 12 - 13 | 1305 | 112 |
AF052059.1 | - | 12 - 13 | 1353 | 112 | |
AK075511.1 | - | 12 - 13 | 1356 | 112 |