The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244984.1 | NP_001231913.1 | 2 - 3 | 223 | 91 |
NM_005065.5 | NP_005056.3 | 2 - 3 | 223 | 91 | |
XM_005267988.2 | XP_005268045.1 | 2 - 3 | 169 | 91 | |
XM_005267989.3 | XP_005268046.1 | 2 - 3 | 226 | 91 | |
GenBank mRNA | AB020335.1 | - | 2 - 3 | 151 | 91 |
AF052059.1 | - | 2 - 3 | 199 | 91 | |
AK075511.1 | - | 2 - 3 | 202 | 91 | |
AY358651.1 | - | 2 - 3 | 195 | 91 | |
BP358857.1 | - | 2 - 3 | 223 | 91 |