The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005065.5 | NP_005056.3 | 8 - 9 | 1006 | 129 |
XM_005267988.2 | XP_005268045.1 | 8 - 9 | 889 | 129 | |
XM_005267989.3 | XP_005268046.1 | 8 - 9 | 1009 | 129 | |
GenBank mRNA | AB020335.1 | - | 8 - 9 | 934 | 129 |
AF052059.1 | - | 8 - 9 | 982 | 129 | |
AK075511.1 | - | 8 - 9 | 985 | 129 |