The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293197.1 | NP_001280126.1 | 2 - 3 | 864 | 77 |
NM_005861.3 | NP_005852.2 | 2 - 3 | 775 | 77 | |
GenBank mRNA | AF039689.1 | - | 2 - 3 | 416 | 77 |
AF129085.1 | - | 2 - 3 | 456 | 77 | |
AF217968.1 | - | 2 - 3 | 751 | 77 | |
AF432221.1 | - | 2 - 3 | 471 | 77 | |
BC007545.1 | - | 2 - 3 | 459 | 77 | |
BC017178.1 | - | 2 - 3 | 446 | 77 | |
BC022788.1 | - | 2 - 3 | 459 | 77 | |
BC063617.1 | - | 2 - 3 | 390 | 77 | |
CB145452.1 | - | 2 - 3 | 284 | 77 |