Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
Chromosome Location
Chr.6: 485138 - 693141 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018303.5 NP_060773.3 10 - 11 1237 71
NR_073064.1 12 - 13 1399 71
XM_017011018.1 XP_016866507.1 11 - 12 1283 71
XM_017011019.1 XP_016866508.1 11 - 12 1445 71
XM_017011020.1 XP_016866509.1 11 - 12 1305 71
XM_017011021.1 XP_016866510.1 10 - 11 1541 71
XM_017011022.1 XP_016866511.1 10 - 11 1237 71
XM_017011023.1 XP_016866512.1 10 - 11 1237 71
XM_017011024.1 XP_016866513.1 11 - 12 1283 71
XM_017011025.1 XP_016866514.1 10 - 11 1237 71
XM_017011026.1 XP_016866515.1 10 - 11 1237 71
GenBank mRNA AJ414403.1 - 10 - 11 1161 71
AK001888.1 - 6 - 7 652 71
AK056564.1 - 10 - 11 1205 71
AK314628.1 - 10 - 11 1237 71
AL833209.1 - 8 - 9 793 71
AL833213.1 - 10 - 11 1183 71
BC016918.1 - 7 - 8 717 71
BC080542.1 - 12 - 13 1369 71

View Full Product Details