Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
Chromosome Location
Chr.6: 485138 - 693141 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018303.5 NP_060773.3 26 - 27 2787 115
NR_073064.1 28 - 29 2949 115
XM_017011018.1 XP_016866507.1 27 - 28 2833 115
XM_017011019.1 XP_016866508.1 27 - 28 2995 115
XM_017011020.1 XP_016866509.1 27 - 28 2855 115
XM_017011021.1 XP_016866510.1 26 - 27 3091 115
XM_017011022.1 XP_016866511.1 26 - 27 2787 115
XM_017011023.1 XP_016866512.1 26 - 27 2787 115
XM_017011024.1 XP_016866513.1 27 - 28 2833 115
GenBank mRNA AJ414403.1 - 26 - 27 2711 115
AJ420556.1 - 4 - 5 306 115
AK001888.1 - 22 - 23 2202 115
AK056564.1 - 26 - 27 2755 115
AK302941.1 - 16 - 17 1528 115
AK314628.1 - 26 - 27 2787 115
AL833209.1 - 24 - 25 2343 115
AL833213.1 - 26 - 27 2733 115
BC016918.1 - 23 - 24 2267 115
BC080542.1 - 28 - 29 2919 115

View Full Product Details