Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOC2
Entrez Gene ID
55770
Gene Name
exocyst complex component 2
Gene Aliases
SEC5, SEC5L1, Sec5p
Chromosome Location
Chr.6: 485138 - 693141 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018303.5 NP_060773.3 7 - 8 912 88
NR_073064.1 9 - 10 1074 88
XM_017011018.1 XP_016866507.1 8 - 9 958 88
XM_017011019.1 XP_016866508.1 8 - 9 1120 88
XM_017011020.1 XP_016866509.1 8 - 9 980 88
XM_017011021.1 XP_016866510.1 7 - 8 1216 88
XM_017011022.1 XP_016866511.1 7 - 8 912 88
XM_017011023.1 XP_016866512.1 7 - 8 912 88
XM_017011024.1 XP_016866513.1 8 - 9 958 88
XM_017011025.1 XP_016866514.1 7 - 8 912 88
XM_017011026.1 XP_016866515.1 7 - 8 912 88
GenBank mRNA AJ414403.1 - 7 - 8 836 88
AK001888.1 - 3 - 4 327 88
AK056564.1 - 7 - 8 880 88
AK314628.1 - 7 - 8 912 88
AL833209.1 - 5 - 6 468 88
AL833213.1 - 7 - 8 858 88
BC016918.1 - 4 - 5 392 88
BC080542.1 - 9 - 10 1044 88

View Full Product Details