The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160328.1 | NP_001153800.1 | 6 - 7 | 1841 | 62 |
NM_001160329.1 | NP_001153801.1 | 6 - 7 | 1477 | 62 | |
NM_032298.2 | NP_115674.1 | 4 - 5 | 1912 | 62 | |
XM_011527390.2 | XP_011525692.1 | 6 - 7 | 2899 | 62 | |
XM_011527391.2 | XP_011525693.1 | 6 - 7 | 1549 | 62 | |
GenBank mRNA | AL136594.1 | - | 4 - 5 | 1912 | 62 |
BC028379.1 | - | 6 - 7 | 1837 | 62 | |
BC031067.1 | - | 6 - 7 | 1460 | 62 |