The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000379.3 | NP_000370.2 | 26 - 27 | 3044 | 77 |
XM_011533095.2 | XP_011531397.1 | 26 - 27 | 3073 | 77 | |
XM_011533096.2 | XP_011531398.1 | 26 - 27 | 3075 | 77 | |
GenBank mRNA | D11456.2 | - | 26 - 27 | 3022 | 77 |
U06117.1 | - | 26 - 27 | 3028 | 77 | |
U39487.1 | - | 26 - 27 | 2999 | 77 |