The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000379.3 | NP_000370.2 | 8 - 9 | 730 | 63 |
XM_011533095.2 | XP_011531397.1 | 8 - 9 | 762 | 63 | |
XM_011533096.2 | XP_011531398.1 | 8 - 9 | 761 | 63 | |
GenBank mRNA | D11456.2 | - | 8 - 9 | 708 | 63 |
U39487.1 | - | 8 - 9 | 685 | 63 |