Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
Chromosome Location
Chr.11: 72223861 - 72239147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001567.3 NP_001558.3 13 - 14 1759 90
XM_005273978.3 XP_005274035.1 14 - 15 2136 90
XM_005273979.4 XP_005274036.1 14 - 15 1983 90
XM_011544999.2 XP_011543301.1 14 - 15 1917 90
XM_011545000.2 XP_011543302.1 14 - 15 1983 90
XM_017017673.1 XP_016873162.1 15 - 16 2074 90
GenBank mRNA BC140853.1 - 13 - 14 1759 90
L36818.1 - 12 - 13 1736 90
Y14385.1 - 13 - 14 1823 90

View Full Product Details