The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001567.3 | NP_001558.3 | 25 - 26 | 3032 | 62 |
XM_005273978.3 | XP_005274035.1 | 26 - 27 | 3409 | 62 | |
XM_005273979.4 | XP_005274036.1 | 26 - 27 | 3256 | 62 | |
XM_011544999.2 | XP_011543301.1 | 26 - 27 | 3190 | 62 | |
XM_011545000.2 | XP_011543302.1 | 26 - 27 | 3256 | 62 | |
XM_017017673.1 | XP_016873162.1 | 27 - 28 | 3347 | 62 | |
GenBank mRNA | BC140853.1 | - | 25 - 26 | 3032 | 62 |
L36818.1 | - | 24 - 25 | 3009 | 62 | |
Y14385.1 | - | 25 - 26 | 3096 | 62 |