Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPGRIP1L
Entrez Gene ID
23322
Gene Name
RPGRIP1 like
Gene Aliases
CORS3, FTM, JBTS7, MKS5, NPHP8, PPP1R134
Chromosome Location
Chr.16: 53599239 - 53703934 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005255867.1 XP_005255924.1 12 - 13 1434 136
XM_005255868.1 XP_005255925.1 13 - 14 1446 136
XM_011522970.1 XP_011521272.1 13 - 14 1446 136
XM_011522971.2 XP_011521273.1 13 - 14 1446 136
XM_011522973.2 XP_011521275.1 13 - 14 1446 136
XM_017023094.1 XP_016878583.1 13 - 14 1446 136
XM_017023095.1 XP_016878584.1 13 - 14 1446 136
XM_017023096.1 XP_016878585.1 13 - 14 1446 136
XM_017023097.1 XP_016878586.1 13 - 14 1446 136
XM_017023100.1 XP_016878589.1 13 - 14 1446 136
GenBank mRNA AB023222.1 - 8 - 9 863 136
AK096333.1 - 12 - 13 1445 136
AK304790.1 - 12 - 13 1496 136
AK308390.1 - 12 - 13 1445 136
BC136433.1 - 12 - 13 1439 136
CR749645.1 - 12 - 13 1453 136

View Full Product Details