The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000704.2 | NP_000695.2 | 11 - 12 | 1727 | 88 |
GenBank mRNA | AL832971.1 | - | 10 - 11 | 1605 | 88 |