Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286164.1 NP_001273093.1 5 - 6 1352 82
NM_004319.2 NP_004310.1 5 - 6 1352 82
NM_207108.2 NP_996991.1 5 - 6 1352 82
XM_017001340.1 XP_016856829.1 5 - 6 1302 82
XM_017001341.1 XP_016856830.1 5 - 6 1302 82
GenBank mRNA AB006627.1 - 5 - 6 1150 82
AK295126.1 - 5 - 6 1304 82
AK307820.1 - 5 - 6 1263 82
AK309094.1 - 5 - 6 1421 82
BC142697.1 - 5 - 6 1131 82
BC146795.1 - 5 - 6 1150 82

View Full Product Details