Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017001340.1 XP_016856829.1 7 - 8 1639 95
XM_017001341.1 XP_016856830.1 7 - 8 1639 95

View Full Product Details