Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASTN1
Entrez Gene ID
460
Gene Name
astrotactin 1
Gene Aliases
ASTN
Chromosome Location
Chr.1: 176855118 - 177164904 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286164.1 NP_001273093.1 8 - 9 1750 73
NM_004319.2 NP_004310.1 8 - 9 1750 73
NM_207108.2 NP_996991.1 8 - 9 1750 73
XM_017001340.1 XP_016856829.1 8 - 9 1724 73
XM_017001341.1 XP_016856830.1 8 - 9 1724 73
GenBank mRNA AB006627.1 - 8 - 9 1548 73
AK295126.1 - 8 - 9 1702 73
AK307820.1 - 8 - 9 1661 73
AK309094.1 - 8 - 9 1819 73
BC142697.1 - 8 - 9 1529 73
BC146795.1 - 8 - 9 1548 73

View Full Product Details