Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE2
Entrez Gene ID
221184
Gene Name
copine 2
Gene Aliases
COPN2, CPN2
Chromosome Location
Chr.16: 57092543 - 57147966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152727.5 NP_689940.3 7 - 8 959 98
GenBank mRNA AF492484.1 - 7 - 8 910 98
AK292269.1 - 6 - 7 868 98
AK315394.1 - 7 - 8 862 98
BC053658.1 - 8 - 9 1033 98
CR749617.1 - 6 - 7 620 98

View Full Product Details