The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007006.2 | NP_008937.1 | 4 - 5 | 614 | 146 |
GenBank mRNA | AJ001810.1 | - | 4 - 5 | 467 | 146 |
AL535307.3 | - | 4 - 5 | 614 | 146 | |
BC001403.2 | - | 4 - 5 | 575 | 146 | |
BX537360.1 | - | 4 - 5 | 618 | 146 | |
CR456919.1 | - | 4 - 5 | 467 | 146 |